Biotin maleimide, ReadiView™ biodetection

Supplier: AAT Bioquest
Warning

3058
76481-644EA 249.58 USD
76481-644
Biotin maleimide, ReadiView™ biodetection
Biotin maleimide

Biotin/avidin complexes are widely applied for a variety of biological detections.


  • High fluorescence enhancement
  • Sensitive detection
  • Multiple applications

Formula: C20H29N5O5S
CAS Number: 116919-18-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71e5f9d03d00ab244907dd197e6c1f56dabba24c&vid=71e5f9d03d00ab244907dd197e6c1f56dabba24c&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d64a665c93ab0c5398a243ae78102f6c179ace8f&vid=9ad4bbcc7d4a6568eeacde5c881952f436fbdd55&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR