Biotin-PEG2-Azide ≥95%

Supplier: AAT Bioquest
Warning

3019
76481-630EA 124.31 USD
76481-630
Biotin-PEG2-Azide ≥95%
Biotin-PEG2-Azide

Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins.

Formula: C₁₆H₂₈N₆O₄S
MDL Number: MFCD26142991
CAS Number: 945633-30-7
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR