Biotin-PEG2-NHS biodetection

Supplier: AAT BIOQUEST INC
Warning

3016
76481-626EA 124.31 USD
76481-626
Biotin-PEG2-NHS biodetection
Biotin-PEG2-NHS

This amine-reactive biotin derivative contains a long arm (~20 Å) to increase its avidin-binding affinity.


  • Improved labeling efficiency
  • High binding affinity

Formula: C₂₁H₃₂N₄O₈S
MDL Number: MFCD00467154
CAS Number: 596820-83-6
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a5f552085b7b8e7dfc1f344ba44f1af75e198ae&vid=bcdda35df7d263e310b43dfe20aeb475e8f51743&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25e79cfe15e454c334675228cac20099ba0724b4&vid=25e79cfe15e454c334675228cac20099ba0724b4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR