D-Biotin-N-succinimidyl ester ≥90%

Supplier: AAT Bioquest
Warning

3002
76481-606EA 249.58 USD
76481-606
D-Biotin-N-succinimidyl ester ≥90%
D-Biotin-N-succinimidyl ester

Biotin succinimidyl ester is the most popular amine-reactive biotin derivative for modifying proteins and other biological molecules.

Formula: C₁₄H₁₉N₃O₅S
MDL Number: MFCD00078531
CAS Number: 35013-72-0
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR