D-(+)-Biotin ≥90%

Supplier: AAT Bioquest
Warning

3001
76481-604EA 96.26 USD
76481-604
D-(+)-Biotin ≥90%
D(+)-Biotin

Biotin binds with high affinity to both avidins and streptavidins.

Formula: C₁₀H₁₆N₂O₃S
MDL Number: MFCD00005541
CAS Number: 58-85-5
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR