Trypan blue, Ultra Pure Grade live cell labeling

Supplier: AAT BIOQUEST INC
Warning

2452
76481-556EA 249.58 USD
76481-556
Trypan blue, Ultra Pure Grade live cell labeling
Trypan Blue

Trypan blue, a diazo dye is a vital stain that colors dead tissues or cells blue.


  • Vital stain
  • High purity

Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number: MFCD00003969
CAS Number: 72-57-1
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6c34e35d5e93c068248117ec70ee42da3b18999&vid=27a391c8e60d5585846d4660f8487803d3cafbb2&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96e443cf06ea5ada2d0cedc67877aa1a0073b147&vid=96e443cf06ea5ada2d0cedc67877aa1a0073b147&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR