Trypan blue ≥75%

Supplier: AAT BIOQUEST INC
Warning

2450
76481-554EA 1195.79 USD
76481-554
Trypan blue ≥75%
Trypan Blue

Trypan blue, a diazo dye is a vital stain that colors dead tissues or cells blue.

Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number: MFCD00003969
CAS Number: 72-57-1
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR