Tide Fluor™ 3WS amine, Tide Fluor™ fluorescent probe

Supplier: AAT Bioquest
Warning

2347
76481-544EA 249.58 USD
76481-544
Tide Fluor™ 3WS amine, Tide Fluor™ fluorescent probe
Tide Fluor™ 3WS amine

Tide Fluor™ 3WS (TF3WS) family has the spectral properties essentially identical to those of Cy3.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a680828403d9e6c981315a272177506ea519af9a&vid=a680828403d9e6c981315a272177506ea519af9a&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb8c122567a4bdd4def75f055df350070cff3327&vid=f0ef295516b74d9bb8c0c326f0fe2e62ba0d0ed6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR