Tide Fluor™ 3WS acid, Tide Fluor™ fluorescent probe

Supplier: AAT Bioquest
Warning

2345
76481-540EA 375.27 USD
76481-540
Tide Fluor™ 3WS acid, Tide Fluor™ fluorescent probe
Tide Fluor™ 3WS acid

Tide Fluor™ 3WS (TF3WS) family has the spectral properties essentially identical to those of Cy3.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b1280632940b2e3b9003fc1faee83bd20e6a42&vid=210f2e7c2c16cf7cffc4b6bb475bee67bbd50a82&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bc0f9ad883316e22229e8d3e7779c09f60ca772&vid=3bc0f9ad883316e22229e8d3e7779c09f60ca772&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR