Tide Fluor™ 7WS maleimide, Tide Fluor™ fluorescent probe

Supplier: AAT Bioquest
Warning

2332
76481-526EA 249.58 USD
76481-526
Tide Fluor™ 7WS maleimide, Tide Fluor™ fluorescent probe
Tide Fluor™ 7WS maleimide

Tide Fluor™ 7WS (TF7WS) family has the spectral properties similar to those of Cy7, IRDye 800 and Alexa Fluor 750.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b8aa9b4baf9ddb77a61b518d1e317a69cf06f4b&vid=9b8aa9b4baf9ddb77a61b518d1e317a69cf06f4b&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1eee688adc7e36ae7f2932e1499a9b783faafbc9&vid=5f9360f8bda7721fa6fc191d6c797f8a618f761b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR