Tide Fluor™ 6WS alkyne, Tide Fluor™ fluorescent probe

Supplier: AAT Bioquest
Warning

2303
76481-512EA 249.58 USD
76481-512
Tide Fluor™ 6WS alkyne, Tide Fluor™ fluorescent probe
Tide Fluor™ 6WS alkyne

Tide Fluor™ 6 (TF6WS) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc384b5641fa2c81a4e376da6442f264c7de805b&vid=312c4a7e5d931aa5ffa592ab88f5e7805a536926&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ffd5c7571abef29003b01cefc1644d17fedc5f6&vid=8ffd5c7571abef29003b01cefc1644d17fedc5f6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR