Tide Fluor™ 6WS azide, Tide Fluor™ fluorescent probe

Supplier: AAT Bioquest
Warning

2302
76481-510EA 249.58 USD
76481-510
Tide Fluor™ 6WS azide, Tide Fluor™ fluorescent probe
Tide Fluor™ 6WS azide

Tide Fluor™ 6WS (TF6WS) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=490868b7782106ab231dd2a852575f02729b618a&vid=490868b7782106ab231dd2a852575f02729b618a&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0b3cbb644631b33457602438d522b1a668ebf49&vid=46bc2316ca71e9f62fb5b447e2da6d2e3e2a17a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR