Tide Fluor™ 4 maleimide, Tide Fluor™ fluorescent probe

Supplier: AAT Bioquest
Warning

2287
76481-494EA 249.58 USD
76481-494
Tide Fluor™ 4 maleimide, Tide Fluor™ fluorescent probe
Tide Fluor™ 4 maleimide

TF4 is designed to be a superior fluorophore alternative to ROX and Texas Red®.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ba690841568f6e56e659ae9f954e9906b6046db&vid=6ba690841568f6e56e659ae9f954e9906b6046db&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c666e63b37a5674fa60aa9db1a00dae458db52e&vid=b9bde6af6b97d3c9456f54c47262285831f4f408&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR