Tide Fluor™ 5WS succinimidyl ester, Tide Fluor™ fluorescent probe

Supplier: AAT Bioquest
Warning

2281
76481-488EA 375.27 USD
76481-488
Tide Fluor™ 5WS succinimidyl ester, Tide Fluor™ fluorescent probe
Tide Fluor™ 5WS succinimidyl ester

Tide Fluor™ 5WS (TF5WS) family has the spectral properties essentially identical to those of Cy5.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc5bb0ef80e85003b2fbe1f29b7afc8773a9f663&vid=1699fbc755cd2fc8436c44fe6e3aa0ffb17d5ad7&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2cb8d0c64ece857a26232a17a11496d50964bac&vid=c2cb8d0c64ece857a26232a17a11496d50964bac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR