Tide Fluor™ 1 CPG 500 Å, Tide Fluor™ fluorescent probe

Supplier: AAT Bioquest
Warning

2240
76481-446EA 249.58 USD
76481-446
Tide Fluor™ 1 CPG 500 Å, Tide Fluor™ fluorescent probe
Tide Fluor™ 1 CPG

TF1 is designed to be a superior fluorophore alternative to EDANS.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86febbd0a2801e773dfa20b7c24e0609c814344e&vid=86febbd0a2801e773dfa20b7c24e0609c814344e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR