Tide Quencher™ 3WS acid, Tide Quencher™ fluorescent probe
Supplier: AAT Bioquest
Warning
2227
76481-426EA 249.58 USD
76481-426
Tide Quencher™ 3WS acid, Tide Quencher™ fluorescent probe
Tide Quencher™ 3WS acid
TQ3WS is designed to be a superior quencher to TAMRA, TF3 and Cy3.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b82edf1fa09e62c999d409dc35679cea288d8ade&vid=cfb16d3273c20842f0f1ba84cb36e460a2fdf41d&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68fd07304df7996a1f5f03f3a5966bf6675afa3d&vid=68fd07304df7996a1f5f03f3a5966bf6675afa3d&prid=web&clid=SAPDAM