Tide Quencher™ 2 alkyne, Tide Quencher™ fluorescent probe
Supplier: AAT Bioquest
Warning
2212
76481-414EA 249.58 USD
76481-414
Tide Quencher™ 2 alkyne, Tide Quencher™ fluorescent probe
Tide Quencher™ 2 alkyne
TQ2 is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f4eaaba435d9f3cfc6110f8db26f66a2893e089&vid=a5e54ca68e61e6a241465b6400bd8d238564ac8e&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85b4d8b4ade90fa04f7a6a2aa17cf841cd041332&vid=85b4d8b4ade90fa04f7a6a2aa17cf841cd041332&prid=web&clid=SAPDAM