Tide Quencher™ 2 azide, Tide Quencher™ fluorescent probe
Supplier: AAT Bioquest
Warning
2211
76481-412EA 249.58 USD
76481-412
Tide Quencher™ 2 azide, Tide Quencher™ fluorescent probe
Tide Quencher™ 2 azide
TQ2 is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=853c246c0931cde31358dda5ab6c8860f7fa46e9&vid=cf9192b1c76c829f974135057f64b0ba9a5d20fc&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a77ac09b2e151fc9b10607d3053b0ca888674870&vid=a77ac09b2e151fc9b10607d3053b0ca888674870&prid=web&clid=SAPDAM