Tide Quencher™ 7WS azide, Tide Quencher™ fluorescent probe
Supplier: AAT Bioquest
Warning
2112
76481-378EA 500.94 USD
76481-378
Tide Quencher™ 7WS azide, Tide Quencher™ fluorescent probe
TQ7WS is designed to be a superior quencher to Cy7, TF7 and Alexa Fluor 750.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97dc8787fa501eda182cdd8f884c12eb7721ff38&vid=af0dd4e8b9299c78038acfaeccad1a2ff6eb42f0&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c6e6111b40f762ed0bf380d3298533602c701a3&vid=1c6e6111b40f762ed0bf380d3298533602c701a3&prid=web&clid=SAPDAM