Tide Quencher™ 7WS acid, Tide Quencher™ fluorescent probe
Supplier: AAT Bioquest
Warning
2105
76481-370EA 626.51 USD
76481-370
Tide Quencher™ 7WS acid, Tide Quencher™ fluorescent probe
Tide Quencher™ 7WS acid
TQ7WS is designed to be a superior quencher to Cy7, TF7 and Alexa Fluor 750.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c3bcd088b0450fe80eea13571c76c2338751708&vid=2c3bcd088b0450fe80eea13571c76c2338751708&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a27f2ee9a12b2103b66a7e14b08623fc255e9f76&vid=b0c107de77e434462426cfbe7d616629e4c6b699&prid=web&clid=SAPDAM