Tide Quencher™ 6WS succinimidyl ester, Tide Quencher™ fluorescent probe
Supplier: AAT Bioquest
Warning
2096
76481-364EA 249.58 USD
76481-364
Tide Quencher™ 6WS succinimidyl ester, Tide Quencher™ fluorescent probe
Tide Quencher™ 6WS succinimidyl ester
TQ6WS is designed to be a superior quencher to Cy5.5, TF6 and Alexa Fluor 680.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97b39f1e6f6f31e965822c9b353769e2a154918f&vid=f9db8900df07bb6e2912a44a4f7da19adda57f34&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0185789b531514db04a0277c4cddcadb692d7826&vid=0185789b531514db04a0277c4cddcadb692d7826&prid=web&clid=SAPDAM