Tide Quencher™ 5 CPG 1000 Å, Tide Quencher™ fluorescent probe
Supplier: AAT Bioquest
Warning
2078
76481-348EA 223.27 USD
76481-348
Tide Quencher™ 5 CPG 1000 Å, Tide Quencher™ fluorescent probe
Tide Quencher™ 5 CPG
TQ5 is designed to be a superior quencher to Cy5, TF5 and Cy5.5.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e615ed397a394a20cb93c96f1fca9f5405f5756&vid=8e615ed397a394a20cb93c96f1fca9f5405f5756&prid=web&clid=SAPDAM