Tide Quencher™ 4WS alkyne, Tide Quencher™ fluorescent probe
Supplier: AAT Bioquest
Warning
2069
76481-340EA 375.27 USD
76481-340
Tide Quencher™ 4WS alkyne, Tide Quencher™ fluorescent probe
TQ4WS is designed to be a superior quencher to ROX, TF4, iFluor® 594, Alexa Fluor® 594 and Texas Red®.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bb5096a77894c834dea3ecd2d092e2ff4f32d39&vid=66efbe97b6f3e7fda7875633f4a5d2ad17e99749&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9999f21795090730a26c24b9fde034615e085580&vid=9999f21795090730a26c24b9fde034615e085580&prid=web&clid=SAPDAM