Tide Quencher™ 4 CPG 500 Å, Tide Quencher™ fluorescent probe
Supplier: AAT Bioquest
Warning
2062
76481-330EA 223.27 USD
76481-330
Tide Quencher™ 4 CPG 500 Å, Tide Quencher™ fluorescent probe
Tide Quencher™ 4 CPG
TQ4 is designed to be a superior quencher to ROX, TF4 and Texas Red®.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80d4ba9672d7a17535d6cf0c9fca82a1a234a274&vid=80d4ba9672d7a17535d6cf0c9fca82a1a234a274&prid=web&clid=SAPDAM