Violet 540 succinimidyl ester, mFluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Violet 540 SE

1152
76481-242EA 249.58 USD
76481-242
Violet 540 succinimidyl ester, mFluor™ fluorescent probe
Violet 540 succinimidyl ester

AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.


  • pH-insensitive
  • High photostability
  • Brighter signal

MW: 1535.88 g/mol
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0662a5afb8adc4a54d3619f8eab8a68dd0d7ed5e&vid=0662a5afb8adc4a54d3619f8eab8a68dd0d7ed5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR