ReadiLink™ Rapid mFluor™ Blue 570 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Supplier: AAT BIOQUEST INC
1120
76481-230EA 188.87 USD
76481-230
ReadiLink™ Rapid mFluor™ Blue 570 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Protein Linking and Labeling Systems
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
  • Blue-laser excitable
  • Robust and simple antibody labeling protocol
  • No column purification required
Caution: For research use only.