ReadiLink™ Rapid mFluor™ Violet 450 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Supplier: AAT BIOQUEST INC
1100
76481-222EA 188.87 USD
76481-222
ReadiLink™ Rapid mFluor™ Violet 450 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* (2 Labelings), AAT Bioquest
Protein Linking and Labeling Systems
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
  • Suitable for Pacific Blue™ channel
  • robust and simple antibody labeling protocol
  • no column purification required
Caution: For research use only.