iFluor® 647, Alkyne
Supplier: AAT BIOQUEST INC
Warning
1090
76481-218EA 249.58 USD
76481-218
iFluor® 647, Alkyne
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d7b0e56dd904641ded5de5d53193d1f9ee66fba&vid=4d7b0e56dd904641ded5de5d53193d1f9ee66fba&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf1a9731f9d6c7e00868249e63fe39a146490f64&vid=48c777ee77931432ac577e27eef53464f54698c6&prid=web&clid=SAPDAM