iFluor® 647, Hydrazide

Supplier: AAT BIOQUEST INC
Warning

1085
76481-210EA 249.58 USD
76481-210
iFluor® 647, Hydrazide
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • pH independent fluorescence between 3-11

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72a269fe1e817f92cb48d71a29997c7cdbbb491c&vid=62013399bb79b5fc86824936efdcfe9e5a7b6b22&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40c3e8bada47cdb84ecb217bd2fc54d250474b3e&vid=40c3e8bada47cdb84ecb217bd2fc54d250474b3e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR