iFluor® 488, Hydrazide

Supplier: AAT BIOQUEST INC
Warning

1082
76481-206EA 249.58 USD
76481-206
iFluor® 488, Hydrazide
iFluor®

Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.


  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • pH independent fluorescence between 4-10

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=943fba91defc5d5036b4cdb234850792021fb3bb&vid=943fba91defc5d5036b4cdb234850792021fb3bb&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ab0653452b6690bd1de93190a2d1bcb3707dae1&vid=eb6f135981ad3ee828776af615a1e635ea2df990&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR