iFluor® 647, Maleimide

Supplier: AAT BIOQUEST INC
Warning

1065
76481-176EA 249.58 USD
76481-176
iFluor® 647, Maleimide
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • pH independent fluorescence between 3-11

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f0104b92dde09a580e2abdc4730e8cf981efdd0&vid=5f0104b92dde09a580e2abdc4730e8cf981efdd0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR