iFluor® 488, Maleimide

Supplier: AAT Bioquest
Warning

1062
76481-172EA 249.58 USD
76481-172
iFluor® 488, Maleimide
iFluor®

Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.


  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • pH independent fluorescence between 4-10

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56b84aac96657b32cf83a695b96d93b76dde2b4&vid=f56b84aac96657b32cf83a695b96d93b76dde2b4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR