Dansyl chloride ≥95%

Supplier: AAT BIOQUEST INC
Warning

811
76481-016EA 96.26 USD
76481-016
Dansyl chloride ≥95%
Dansyl chloride

Dansyl chloride is non-fluorescent until it reacts with amines.

Formula: C₁₂H₁₂ClNO₂S
MDL Number: MFCD00003985
CAS Number: 605-65-2
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR