2-Aminoacridone ≥95%

Supplier: AAT Bioquest
Warning

604
76480-990EA 96.26 USD
76480-990
2-Aminoacridone ≥95%
2-Aminoacridone

This reagent is used to modify carbonyl-containing biomolecules, in particular, for labeling carbohydrates and glycoproteins.

Formula: C₁₃H₁₀N₂O
MDL Number: MFCD00037397
CAS Number: 27918-14-5
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR