N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate ≥90%

Supplier: AAT BIOQUEST INC
Warning

551
76480-970EA 124.31 USD
76480-970
N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate ≥90%
N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate

7-Hydroxycoumarin-3-carboxylic acid, SE is the amine-reactive succinimidyl ester of 7-Hydroxycoumarin-3-carboxylic acid.

Formula: C₁₄H₉NO₇
MDL Number: MFCD00037568
CAS Number: 134471-24-2
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR