Tetramethylrhodamine-6-maleimide ≥90%

Supplier: AAT Bioquest
Warning

419
76480-892EA 249.58 USD
76480-892
Tetramethylrhodamine-6-maleimide ≥90%
Formula: C₂₈H₂₃N₃O₅
CAS Number: 174568-68-4
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR