Sunnyvale Red/trade; SE Superior 6-ROX Replacement, Although ROX dyes have strong fluorescence they are notoriously unstable. Some commercial samples only have a few months shelf life, Size: 5mg
Supplier: AAT Bioquest
Sunnyvale Red/trade; SE Superior 6-ROX Replacement, Although ROX dyes have strong fluorescence they are notoriously unstable. Some commercial samples only have a few months shelf life, Size: 5mg