6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE) ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 6-ROX, SE, 6-Carboxy-X-rhodamine, NHS ester, 6-ROX N-succinimidyl ester, Carboxy-X-rhodamine succinimidyl ester, 6-isomer

392 399
76480-868EA 124.31 USD
76480-868 76480-870
6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE) ≥90%
6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE)

6-ROX, SE is the other purified single isomer of 5(6)-ROX, SE.

Formula: C₃₇H₃₃N₃O₇
MW: 631.685 g/mol
CAS Number: 216699-36-4
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3adeec2e618d117fc43365188bfd472486b11903&vid=240025677973921ee697127568bed3c40f01c362&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR