5-Carboxytetramethylrhodamine (5-TAMRA) ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer
363
364
76480-824EA
96.26
USD
76480-824
76480-826
5-Carboxytetramethylrhodamine (5-TAMRA) ≥95%
5-Carboxytetramethylrhodamine (5-TAMRA)
5-TAMRA is the purified single isomer of 5(6)-TAMRA.
Formula:
C₂₅H₂₂N₂O₅ |
MDL Number:
MFCD00269769 CAS Number: 91809-66-4 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...