5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF) mixture of isomers ≥95%

Supplier: AAT BIOQUEST INC

Synonyms: Carboxy-DCF, 5(6)-CDCF

235
76480-750EA 124.31 USD
76480-750
5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF) mixture of isomers ≥95%
5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF)

CDCF [5(6)-carboxy-2',7'-dichlorofluorescein] is the fluorescent product of 5(6)-carboxy-2',7'-dichlorodihydrofluorescein diacetate and 5(6)-carboxy-2',7'-dichlorofluorescein diacetate in cells.


  • ROS Indicator
  • Stable Fluorescence


Caution: For Research Use Only

Formula: C₂₁H₁₀Cl₂O₇
MDL Number: MFCD00037452
CAS Number: 111843-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR