Fluorescein-5-maleimide ≥90%

Supplier: AAT Bioquest
Warning

130
76480-598EA 159.77 USD
76480-598
Fluorescein-5-maleimide ≥90%
Fluorescein-5-maleimide
Formula: C₂₄H₁₃NO₇
MDL Number: MFCD00077345
CAS Number: 75350-46-8
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR