6-Carboxyfluorescein ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM

106 107 108
76480-554EA 96.26 USD
76480-554 76480-556 76480-558
6-Carboxyfluorescein ≥95%
6-Carboxyfluorescein

6-FAM is the other purified isomer of 5(6)-carboxyfluorescein.

Formula: C₂₁H₁₂O₇
MW: 376.32 g/mol
MDL Number: MFCD00036873
CAS Number: 3301-79-9
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd80d3ff1bb5b71ce65a794dee606f3900284a92&vid=3ab92665e6217afe078170a638027f190dd477bb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR