Bromocresol purple ≥95%, Ultra Pure Grade

Supplier: AAT Bioquest
Warning

92
76480-538EA 375.27 USD
76480-538
Bromocresol purple ≥95%, Ultra Pure Grade
Bromocresol purple

Bromocresol purple (BCP) or 5',5-dibromo-o-cresolsulfophthalein, is a pH indicator.

Formula: C₂₁H₁₆Br₂O₅S
MDL Number: MFCD00011681
CAS Number: 115-40-2
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR