7-Hydroxy-4-(trifluoromethyl)coumarin

Supplier: AAT BIOQUEST INC
Warning

82
76480-524EA 96.26 USD
76480-524
7-Hydroxy-4-(trifluoromethyl)coumarin
7-Hydroxy-4-(trifluoromethyl)coumarin

This coumarin compound is used as a calibration standard for 7-hydroxy-4-trifluoromethylcoumarin-based enzyme substrates.

Formula: C₁₀H₅F₃O₃
MDL Number: MFCD00037578
CAS Number: 575-03-1
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR