Rhodamine 6G ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

70
76480-512EA 96.26 USD
76480-512
Rhodamine 6G ≥90%
Rhodamine 6G

Rhodamine 6G is also called Rhodamine 590, R6G, Basic Rhodamine Yellow, or C.I.

Formula: C₂₈H₃₁ClN₂O₃
MDL Number: MFCD00012665
CAS Number: 989-38-8
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR