Rhodamine B ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

69
76480-510EA 96.26 USD
76480-510
Rhodamine B ≥95%
Rhodamine B

Rhodamine B (also called Rhodamine 610, Basic Violet 10, or C.I.

Formula: C₂₈H₃₁ClN₂O₃
MDL Number: MFCD00011931
CAS Number: 81-88-9
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR