DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] ≥95%

Supplier: AAT BIOQUEST INC
Warning

62
76480-502EA 124.31 USD
76480-502
DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] ≥95%
DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)]

DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] is used as a calibration standard for DDAO-based enzyme substrates.

Formula: C₁₅H₁₁Cl₂NO₂
MDL Number: MFCD01311087
CAS Number: 118290-05-4
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR