Pyronin G ≥95%

Supplier: AAT BIOQUEST INC
Warning

61
76480-500EA 96.26 USD
76480-500
Pyronin G ≥95%
Pyronine G

Pyronin Y is widely used in combination with methyl green to selectively and differentially stain nucleic acids following polyacrylamide gel electrophoresis.

Formula: C₁₇H₁₉ClN₂O
MDL Number: MFCD00011725
CAS Number: 92-32-0
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR