7-Amino-4-methylcoumarin ≥90%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

51 52 53
76480-486EA 96.26 USD
76480-486 76480-488 76480-490
7-Amino-4-methylcoumarin ≥90%
7-Amino-4-methylcoumarin

This compound is the precursor for a variety of AMC enzyme substrates.

Formula: C₁₀H₉NO₂
MDL Number: MFCD00006868
CAS Number: 26093-31-2
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7232a90dc4e2e8ed29c78554b52f5d0d649282ae&vid=aae157790e6d58d2607c526391a8df661c973e39&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR