4-Methylumbelliferone fluorescence reference standard

Supplier: AAT BIOQUEST INC
Warning

41
76480-482EA 96.26 USD
76480-482
4-Methylumbelliferone fluorescence reference standard
4-Methylumbelliferone

This compound is used as a calibration standard for 7-hydroxy-4-methylcoumarin-based enzyme substrates.


  • pH-dependent fluorescence

Formula: C₁₀H₈O₃
MDL Number: MFCD00006866
CAS Number: 90-33-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64e91c9b42debf9a737d6f1483d1daba47020b70&vid=77bd39318f1a0a01ff57eb9b8df0b8c55e50a109&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=396e4586a2069d357cec19a52f0707554beb100c&vid=396e4586a2069d357cec19a52f0707554beb100c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR